Anti-CD47 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 472603]

Supplier: R & D Systems
FAB4670A
220017-793EA 527.85 USD
220017-793
Anti-CD47 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 472603]
Antibodies
Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: 472603
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR