Anti-SLAMF7 Sheep Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB4628A
220017-789EA 566.89 USD
220017-789
Anti-SLAMF7 Sheep Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: SLAMF7
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR