Anti-SLAMF7 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 520914]

Supplier: R & D Systems
FAB46281P
102882-740EA 513.38 USD
102882-740
Anti-SLAMF7 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 520914]
Antibodies
Type: Primary
Antigen: SLAMF7
Clonality: Monoclonal
Clone: 520914
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR