Anti-CD37 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 424925]

Supplier: R & D Systems
FAB4625P
220017-788EA 561.11 USD
220017-788
Anti-CD37 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 424925]
Antibodies
Type: Primary
Antigen: CD37
Clonality: Monoclonal
Clone: 424925
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR