Anti-CD53 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 425514]

Supplier: R & D Systems
FAB4624A
220017-783EA 513.38 USD
220017-783
Anti-CD53 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 425514]
Antibodies
Type: Primary
Antigen: CD53
Clonality: Monoclonal
Clone: 425514
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR