Anti-CLDN3 Mouse Monoclonal Antibody (Fluorescein) [clone: 385021]

Supplier: R & D Systems
FAB4620F
220017-781EA 420.83 USD
220017-781
Anti-CLDN3 Mouse Monoclonal Antibody (Fluorescein) [clone: 385021]
Antibodies
Type: Primary
Antigen: CLDN3
Clonality: Monoclonal
Clone: 385021
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR