Anti-CLDN3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 385021]

Supplier: R & D Systems
FAB4620A
220017-780EA 608.83 USD
220017-780
Anti-CLDN3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 385021]
Antibodies
Type: Primary
Antigen: CLDN3
Clonality: Monoclonal
Clone: 385021
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR