Anti-CD82 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 423524]

Supplier: R & D Systems
FAB4616A
220017-779EA 543.75 USD
220017-779
Anti-CD82 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 423524]
Antibodies
Type: Primary
Antigen: CD82
Clonality: Monoclonal
Clone: 423524
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR