Anti-CD151 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 455807]

Supplier: R & D Systems
FAB4609P
220017-772EA 561.11 USD
220017-772
Anti-CD151 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 455807]
Antibodies
Type: Primary
Antigen: CD151
Clonality: Monoclonal
Clone: 455807
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR