Anti-CD151 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 455807]

Supplier: R & D Systems
FAB4609A
220017-771EA 543.75 USD
220017-771
Anti-CD151 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 455807]
Antibodies
Type: Primary
Antigen: CD151
Clonality: Monoclonal
Clone: 455807
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR