Anti-PTK7 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 525222]

Supplier: R & D Systems
FAB4499G
102882-712EA 582.8 USD
102882-712
Anti-PTK7 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 525222]
Antibodies
Type: Primary
Antigen: PTK7
Clonality: Monoclonal
Clone: 525222
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR