Anti-NT5E Rat Monoclonal Antibody (Fluorescein) [clone: 496406]

Supplier: R & D Systems
FAB4488F
220017-767EA 349.97 USD
220017-767
Anti-NT5E Rat Monoclonal Antibody (Fluorescein) [clone: 496406]
Antibodies
Type: Primary
Antigen: NT5E
Clonality: Monoclonal
Clone: 496406
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR