Anti-IL17RA Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 657603]

Supplier: R & D Systems
FAB4482A
102882-706EA 577.02 USD
102882-706
Anti-IL17RA Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 657603]
Antibodies
Type: Primary
Antigen: IL17RA
Clonality: Monoclonal
Clone: 657603
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR