Anti-CD40 Rat Monoclonal Antibody (Fluorescein) [clone: 1C10]

Supplier: R & D Systems
FAB440F
220017-762EA 268.98 USD
220017-762
Anti-CD40 Rat Monoclonal Antibody (Fluorescein) [clone: 1C10]
Antibodies
Type: Primary
Antigen: CD40
Clonality: Monoclonal
Clone: 1C10
Conjugation: Fluorescein
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR