Anti-ENTHD1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 498403]

Supplier: R & D Systems
FAB4397A
220017-753EA 501.82 USD
220017-753
Anti-ENTHD1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 498403]
Antibodies
Type: Primary
Antigen: ENTHD1
Clonality: Monoclonal
Clone: 498403
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR