Anti-CD109 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 496920]

Supplier: R & D Systems
FAB4385P
220017-751EA 513.38 USD
220017-751
Anti-CD109 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 496920]
Antibodies
Type: Primary
Antigen: CD109
Clonality: Monoclonal
Clone: 496920
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR