Anti-PILRA Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB4318G
102882-690EA 527.85 USD
102882-690
Anti-PILRA Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: PILRA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR