Anti-DLL3 Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB4315P
102882-688EA 549.54 USD
102882-688
Anti-DLL3 Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: DLL3
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR