Anti-IL22RA1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 496514]

Supplier: R & D Systems
FAB42941P
220017-743EA 513.38 USD
220017-743
Anti-IL22RA1 Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 496514]
Antibodies
Type: Primary
Antigen: IL22RA1
Clonality: Monoclonal
Clone: 496514
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR