Anti-GPBAR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 409522]

Supplier: R & D Systems
FAB4286A
220017-740EA 608.83 USD
220017-740
Anti-GPBAR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 409522]
Antibodies
Type: Primary
Antigen Symbol: GPBAR1
Antigen Name: G protein-coupled bile acid receptor 1
Antigen Synonyms: BG37, GPCR19, GPR131, M-BAR, TGR5
Clonality: Monoclonal
Clone: 409522
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR