Anti-TNFRSF1B Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: TR7554]

Supplier: R & D Systems
FAB426A
220017-738EA 561.11 USD
220017-738
Anti-TNFRSF1B Hamster Monoclonal Antibody (APC (Allophycocyanin)) [clone: TR7554]
Antibodies
Type: Primary
Antigen: TNFRSF1B
Clonality: Monoclonal
Clone: TR7554
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR