Anti-FOLH1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 460420]

Supplier: R & D Systems
FAB4234G
102882-666EA 561.11 USD
102882-666
Anti-FOLH1 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 460420]
Antibodies
Type: Primary
Antigen Symbol: FOLH1
Antigen Name: folate hydrolase (prostate-specific membrane antigen) 1
Antigen Synonyms: FGCP, FOLH, GCP2, GCPII, NAALAD1, NAALAdase, PSM, PSMA, mGCP
Clonality: Monoclonal
Clone: 460420
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR