Anti-ACKR3 Mouse Monoclonal Antibody (Fluorescein) [clone: 358426]

Supplier: R & D Systems
FAB42271F-025 FAB42271F-100
102882-654EA 227.05 USD
102882-654 102882-656
Anti-ACKR3 Mouse Monoclonal Antibody (Fluorescein) [clone: 358426]
Antibodies
Type: Primary
Antigen: ACKR3
Clonality: Monoclonal
Clone: 358426
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR