Anti-ACKR3 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 358426]

Supplier: R & D Systems
FAB42271C-025 FAB42271C-100
102882-650EA 241.51 USD
102882-650 102882-652
Anti-ACKR3 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)) [clone: 358426]
Antibodies
Type: Primary
Antigen: ACKR3
Clonality: Monoclonal
Clone: 358426
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR