Anti-MS4A1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 396444]

Supplier: R & D Systems
FAB4225P
220017-732EA 323.94 USD
220017-732
Anti-MS4A1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 396444]
Antibodies
Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: 396444
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR