Anti-SSTR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 402038]

Supplier: R & D Systems
FAB4224A
220017-728EA 600.15 USD
220017-728
Anti-SSTR2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 402038]
Antibodies
Type: Primary
Antigen: SSTR2
Clonality: Monoclonal
Clone: 402038
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR