Anti-CLDN4 Mouse Monoclonal Antibody (Fluorescein) [clone: 382321]

Supplier: R & D Systems
FAB4219F
220017-724EA 420.83 USD
220017-724
Anti-CLDN4 Mouse Monoclonal Antibody (Fluorescein) [clone: 382321]
Antibodies
Type: Primary
Antigen: CLDN4
Clonality: Monoclonal
Clone: 382321
Conjugation: Fluorescein
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR