Anti-CLDN4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 382321]

Supplier: R & D Systems
FAB4219A
220017-723EA 577.02 USD
220017-723
Anti-CLDN4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 382321]
Antibodies
Type: Primary
Antigen: CLDN4
Clonality: Monoclonal
Clone: 382321
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR