Anti-ESAM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 408519]

Supplier: R & D Systems
FAB4204A
220017-720EA 594.37 USD
220017-720
Anti-ESAM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 408519]
Antibodies
Type: Primary
Antigen: ESAM
Clonality: Monoclonal
Clone: 408519
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR