Anti-PILRB Sheep Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB4189P
102882-644EA 527.85 USD
102882-644
Anti-PILRB Sheep Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: PILRB
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR