Anti-PLXND1 Goat Polyclonal Antibody (Alexa Fluor® 488)

Supplier: R & D Systems
FAB4160G
102882-638EA 535.08 USD
102882-638
Anti-PLXND1 Goat Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Type: Primary
Antigen: PLXND1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR