Anti-ITGB6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 437211]

Supplier: R & D Systems
FAB4155A
220017-718EA 594.37 USD
220017-718
Anti-ITGB6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 437211]
Antibodies
Type: Primary
Antigen: ITGB6
Clonality: Monoclonal
Clone: 437211
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR