Anti-ACKR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 35837]

Supplier: R & D Systems
FAB4139A
220017-715EA 608.83 USD
220017-715
Anti-ACKR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 35837]
Antibodies
Type: Primary
Antigen: ACKR1
Clonality: Monoclonal
Clone: 35837
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR