Anti-CEACAM5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 487609]

Supplier: R & D Systems
FAB41281P
102882-634EA 614.62 USD
102882-634
Anti-CEACAM5 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 487609]
Antibodies
Type: Primary
Antigen: CEACAM5
Clonality: Monoclonal
Clone: 487609
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR