Anti-GP1BA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 486805]

Supplier: R & D Systems
FAB4067A
102882-626EA 561.11 USD
102882-626
Anti-GP1BA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 486805]
Antibodies
Type: Primary
Antigen Symbol: GP1BA
Antigen Name: glycoprotein Ib (platelet), alpha polypeptide
Antigen Synonyms: BDPLT1, BDPLT3, BSS, CD42B, CD42b-alpha, DBPLT3, GP1B, GPIbA, VWDP
Clonality: Monoclonal
Clone: 486805
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR