Anti-ITGB4 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 422325]

Supplier: R & D Systems
FAB4060G
102882-624EA 513.38 USD
102882-624
Anti-ITGB4 Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 422325]
Antibodies
Type: Primary
Antigen: ITGB4
Clonality: Monoclonal
Clone: 422325
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR