Anti-ITGB4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 422325]

Supplier: R & D Systems
FAB4060A
102882-622EA 543.75 USD
102882-622
Anti-ITGB4 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 422325]
Antibodies
Type: Primary
Antigen: ITGB4
Clonality: Monoclonal
Clone: 422325
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR