Anti-F2RL1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 344222]

Supplier: R & D Systems
FAB3949P
220017-703EA 566.89 USD
220017-703
Anti-F2RL1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 344222]
Antibodies
Type: Primary
Antigen: F2RL1
Clonality: Monoclonal
Clone: 344222
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR