Anti-THBD Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 501733]

Supplier: R & D Systems
FAB3947A
220017-700EA 577.02 USD
220017-700
Anti-THBD Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 501733]
Antibodies
Type: Primary
Antigen: THBD
Clonality: Monoclonal
Clone: 501733
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR