Anti-CEACAM6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 439424]

Supplier: R & D Systems
FAB3934A
220017-695EA 642.09 USD
220017-695
Anti-CEACAM6 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 439424]
Antibodies
Type: Primary
Antigen: CEACAM6
Clonality: Monoclonal
Clone: 439424
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR