Anti-CD99 Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB3905P
102882-618EA 480.12 USD
102882-618
Anti-CD99 Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: CD99
Clonality: Polyclonal
Clone: 0
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR