Anti-THBD Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 461714]

Supplier: R & D Systems
FAB3894P
220017-687EA 471.45 USD
220017-687
Anti-THBD Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: 461714]
Antibodies
Type: Primary
Antigen: THBD
Clonality: Monoclonal
Clone: 461714
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR