Anti-THBD Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 461714]

Supplier: R & D Systems
FAB3894A
220017-685EA 561.11 USD
220017-685
Anti-THBD Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 461714]
Antibodies
Type: Primary
Antigen: THBD
Clonality: Monoclonal
Clone: 461714
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR