Anti-CTLA4 Goat Polyclonal Antibody (PE (Phycoerythrin))

Supplier: R & D Systems
FAB386P
220017-680EA 527.85 USD
220017-680
Anti-CTLA4 Goat Polyclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: CTLA4
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR