Anti-CTLA4 Goat Polyclonal Antibody (APC (Allophycocyanin))

Supplier: R & D Systems
FAB386A
102882-614EA 584.25 USD
102882-614
Anti-CTLA4 Goat Polyclonal Antibody (APC (Allophycocyanin))
Antibodies
Type: Primary
Antigen: CTLA4
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR