Anti-PVRL2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 829038]

Supplier: R & D Systems
FAB3869A
102882-608EA 577.02 USD
102882-608
Anti-PVRL2 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 829038]
Antibodies
Type: Primary
Antigen: PVRL2
Clonality: Monoclonal
Clone: 829038
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR