Anti-F2R Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 731115]

Supplier: R & D Systems
FAB3855G
102882-604EA 566.89 USD
102882-604
Anti-F2R Mouse Monoclonal Antibody (Alexa Fluor® 488) [clone: 731115]
Antibodies
Type: Primary
Antigen: F2R
Clonality: Monoclonal
Clone: 731115
Conjugation: Alexa Fluor® 488
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR