Anti-CNR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 368302]

Supplier: R & D Systems
FAB3834P
102882-600EA 561.11 USD
102882-600
Anti-CNR1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 368302]
Antibodies
Type: Primary
Antigen: CNR1
Clonality: Monoclonal
Clone: 368302
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR