Anti-CD27 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 57703]

Supplier: R & D Systems
FAB382P
220017-675EA 339.85 USD
220017-675
Anti-CD27 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 57703]
Antibodies
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: 57703
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR