Anti-CD27 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 57703]

Supplier: R & D Systems
FAB382A
220017-673EA 485.91 USD
220017-673
Anti-CD27 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 57703]
Antibodies
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: 57703
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR